(2S)-2-(tert-Butoxymethyl)oxirane 97%

Lieferant: Apollo Scientific

OR480705-25G
APOSOR480705-25GEA 665 EUR
APOSOR480705-25G
(2S)-2-(tert-Butoxymethyl)oxirane 97%
(2S)-2-(tert-Butoxymethyl)oxirane
Formel: C₇H₁₄O₂
Molecular Weight: 130,185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09750970
CAS-Nummer: 130232-97-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR