Isocytosin 99%

Lieferant: Apollo Scientific

OR480710-100G
APOSOR480710-100GEA 172 EUR
APOSOR480710-100G
Isocytosin 99%
Isocytosine
Formel: C₄H₅N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD00057557
CAS-Nummer: 108-53-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR