2-Amino-5-phenylpyridin 97%

Lieferant: Apollo Scientific

Synonyme: 5-Phenyl-2-pyridylamin, 5-Phenylpyridin-2-amin

OR480727-5G
APOSOR480727-5GEA 71.2 EUR
APOSOR480727-5G
2-Amino-5-phenylpyridin 97%
2-Amino-5-phenylpyridine
Formel: C₁₁H₁₀N₂
Molecular Weight: 170.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01692452
CAS-Nummer: 33421-40-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR