6-Chlorpyrimidin-4,5-diamin 95%
Lieferant: Apollo Scientific
Synonyme:
4,5-Diamino-6-chloropyrimidine, 6-Chloro-4,5-diaminopyrimidine
OR480730-5G
APOSOR480730-5GEA
106
EUR
APOSOR480730-5G
6-Chlorpyrimidin-4,5-diamin 95%
6-Chlorpyrimidin-4,5-diamin
Formel:
C₄H₅ClN₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00023270 CAS-Nummer: 4316-98-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...