2-Chloro-1-methyl-4-nitro-1H-imidazole 95%

Lieferant: Apollo Scientific

OR480731-5G
APOSOR480731-5GEA 1300 EUR
APOSOR480731-5G
2-Chloro-1-methyl-4-nitro-1H-imidazole 95%
2-Chloro-1-methyl-4-nitro-1H-imidazole
Formel: C₄H₄ClN₃O₂
Molecular Weight: 161,546 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01721662
CAS-Nummer: 63634-21-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR