2-Chloro-5-pyrimidinecarbonyl chloride 95%

Lieferant: Apollo Scientific
Gefahr

OR480739-1G
APOSOR480739-1GEA 0 EUR
APOSOR480739-1G
2-Chloro-5-pyrimidinecarbonyl chloride 95%
2-Chloro-5-pyrimidinecarbonyl chloride
Formel: C₅H₂Cl₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09863165
CAS-Nummer: 110099-99-5
UN: 1760
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR