Tetrahydropyran-4,4-dicarbonitrile 95%

Lieferant: Apollo Scientific

OR480742-1G OR480742-5G
APOSOR480742-1GEA 221 EUR
APOSOR480742-1G APOSOR480742-5G
Tetrahydropyran-4,4-dicarbonitrile 95%
Tetrahydropyran-4,4-dicarbonitrile
Formel: C₇H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD19313175
CAS-Nummer: 111511-90-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR