3-(Bromomethyl)benzoic acid 95%
Lieferant: Apollo Scientific
OR480743-25G
APOSOR480743-25GEA
105
EUR
APOSOR480743-25G
3-(Bromomethyl)benzoic acid 95%
3-(Bromomethyl)benzoic acid
Formel:
C8H7BrO2 Molecular Weight: 215.0440 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00045839 CAS-Nummer: 6515-58-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...