Boc-Lys(Fmoc)-OH 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-epsilon-FMOC-N-alpha-BOC-L-Lysine

OR480747-25G
APOSOR480747-25GEA 71.2 EUR
APOSOR480747-25G
Boc-Lys(Fmoc)-OH 97%
Boc-Lys(Fmoc)-OH
Formel: C₂₆H₃₂N₂O₆
Molecular Weight: 468.55 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00062032
CAS-Nummer: 84624-27-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR