2-Brom-5-hydroxybenzonitril 95%

Lieferant: Apollo Scientific
Achtung

OR480751-25G
APOSOR480751-25GEA 157 EUR
APOSOR480751-25G
2-Brom-5-hydroxybenzonitril 95%
2-Brom-5-hydroxybenzonitril
Formel: C₇H₄BrNO
Molecular Weight: 198.0200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08061922
CAS-Nummer: 189680-06-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR