4,6-Dimethylisoxazolo[5,4-b]pyridin-3-ylamine 97+%

Lieferant: Apollo Scientific

Synonyme: 4,6-Dimethylisoxazolo[5,4-b]pyridin-3-amine

OR480757-1G
APOSOR480757-1GEA 391 EUR
APOSOR480757-1G
4,6-Dimethylisoxazolo[5,4-b]pyridin-3-ylamine 97+%
4,6-Dimethylisoxazolo[5,4-b]pyridin-3-ylamine
Formel: C₈H₉N₃O
Molecular Weight: 163,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08060940
CAS-Nummer: 916792-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR