3-Amino-4-(4-methylphenyl)pyrrolidine, 3-BOC protected hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl N-[4-(4-methylphenyl)pyrrolidin-3-yl]carbamate hydrochloride, tert-Butyl (4-(p-tolyl)pyrrolidin-3-yl)carbamate hydrochloride
OR48076-100MG
APOSOR48076-100MGEA
101
EUR
APOSOR48076-100MG
3-Amino-4-(4-methylphenyl)pyrrolidine, 3-BOC protected hydrochloride
3-Amino-4-(4-methylphenyl)pyrrolidine, 3-BOC protected hydrochloride
Formel:
C₁₆H₂₄N₂O₂·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD32111679 CAS-Nummer: 2367002-73-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...