2,4-Diaminopyrimidine-5-carbonitrile 95%

Lieferant: Apollo Scientific

OR480761-5G
APOSOR480761-5GEA 106 EUR
APOSOR480761-5G
2,4-Diaminopyrimidine-5-carbonitrile 95%
2,4-Diaminopyrimidine-5-carbonitrile
Formel: C₅H₅N₅
Lagertemperatur: Raumtemperatur
MDL: MFCD00085600
CAS-Nummer: 16462-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR