Methyl-1-boc-3-pyrrolidinecarboxylate 97%

Lieferant: Apollo Scientific

Synonyme: Pyrrolidine-1,3-dicarboxylic acid 1-tert-Butyl 3-Methyl ester, 1-Boc-3-pyrrolidinecarboxylic acid methyll ester, 1-tert-Butyl 3-Methyl Pyrrolidine-1,3-dicarboxylate, 1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylic acid methyll ester, Methyl 1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylate, Methyl 1-Boc-3-pyrrolidinecarboxylate

OR480762-25G
APOSOR480762-25GEA 75.4 EUR
APOSOR480762-25G
Methyl-1-boc-3-pyrrolidinecarboxylate 97%
Methyl-1-boc-3-pyrrolidinecarboxylate
Formel: C₁₁H₁₉NO₄
Molecular Weight: 229.28 g/mol
Siedepunkt: 105 °C/0.2mmHg
Dichte: 1.08 g/cm³ (25 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD04038683
CAS-Nummer: 122684-33-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR