5-Nitro-1,2-benzisoxazol 97%

Lieferant: Apollo Scientific

OR480767-1G
APOSOR480767-1GEA 119 EUR
APOSOR480767-1G
5-Nitro-1,2-benzisoxazol 97%
5-Nitro-1,2-benzisoxazole
Formel: C₇H₄N₂O₃
Lagertemperatur: Kühlschrank
MDL: MFCD06659648
CAS-Nummer: 39835-28-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR