Methyl-2-aminopyrimidine-5-carboxylate 96%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-aminopyrimidine-5-carboxylate

OR480769-5G
APOSOR480769-5GEA 120 EUR
APOSOR480769-5G
Methyl-2-aminopyrimidine-5-carboxylate 96%
Methyl-2-aminopyrimidine-5-carboxylate
Formel: C₆H₇N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08275693
CAS-Nummer: 308348-93-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR