4-Chloronicotinonitrile 95%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-3-cyanopyridine

OR480773-5G
APOSOR480773-5GEA 208 EUR
APOSOR480773-5G
4-Chloronicotinonitrile 95%
4-Chloronicotinonitrile
Formel: C₆H₃ClN₂
Molecular Weight: 138,5544 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08234759
CAS-Nummer: 89284-61-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR