Pyrazolo[1,5-a]pyridine-7-carbaldehyde 97%

Lieferant: Apollo Scientific

OR480774-1G OR480774-5G
APOSOR480774-1GEA 215 EUR
APOSOR480774-1G APOSOR480774-5G
Pyrazolo[1,5-a]pyridine-7-carbaldehyde 97%
Pyrazolo[1,5-a]pyridine-7-carbaldehyde
Formel: C₈H₆N₂O
Molecular Weight: 146.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06739029
CAS-Nummer: 362661-83-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR