Pyrazolo[1,5-a]pyridine-7-carbaldehyde 97%
Lieferant: Apollo Scientific
OR480774-1G
OR480774-5G
APOSOR480774-1GEA
215
EUR
APOSOR480774-1G
APOSOR480774-5G
Pyrazolo[1,5-a]pyridine-7-carbaldehyde 97%
Pyrazolo[1,5-a]pyridine-7-carbaldehyde
Formel:
C₈H₆N₂O Molecular Weight: 146.15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06739029 CAS-Nummer: 362661-83-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...