3-Acetylbenzolsulfonylchlorid 96%
Lieferant: Apollo Scientific
Synonyme:
3-Acetylbenzol-1-sulfonylchlorid, 3-Acetylbenzolsulfochlorid, 3-Acetylbenzolsulfonsäurechlorid
OR480786-5G
APOSOR480786-5GEA
271
EUR
APOSOR480786-5G
3-Acetylbenzolsulfonylchlorid 96%
3-Acetylbenzolsulfonylchlorid
Formel:
C₈H₇ClO₃S Molecular Weight: 218.66 Lagertemperatur: Raumtemperatur |
MDL:
MFCD03424979 CAS-Nummer: 73035-16-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...