Methyl 5-chloro-4-nitro-thiophene-2-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: 5-Chloro-4-nitrothiophene-2-carboxylic acid methyl ester

OR480790-5G
APOSOR480790-5GEA 119 EUR
APOSOR480790-5G
Methyl 5-chloro-4-nitro-thiophene-2-carboxylate 95%
Methyl 5-chloro-4-nitro-thiophene-2-carboxylate
Formel: C₆H₄ClNO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD08275718
CAS-Nummer: 57800-76-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR