4-Carboxy-3-nitrophenylboronic acid 95%
Lieferant: Apollo Scientific
OR480791-1G
OR480791-5G
APOSOR480791-1GEA
108
EUR
APOSOR480791-1G
APOSOR480791-5G
4-Carboxy-3-nitrophenylboronic acid 95%
4-Carboxy-3-nitrophenylboronic acid
Formel:
C₇H₆BNO₆ Lagertemperatur: Raumtemperatur |
MDL:
MFCD10696661 CAS-Nummer: 80500-28-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...