4-Carboxy-3-nitrophenylboronic acid 95%

Lieferant: Apollo Scientific

OR480791-1G OR480791-5G
APOSOR480791-1GEA 108 EUR
APOSOR480791-1G APOSOR480791-5G
4-Carboxy-3-nitrophenylboronic acid 95%
4-Carboxy-3-nitrophenylboronic acid
Formel: C₇H₆BNO₆
Lagertemperatur: Raumtemperatur
MDL: MFCD10696661
CAS-Nummer: 80500-28-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR