4-Bromo-3-methylthiophene-2-carboxaldehyde 97%

Lieferant: Apollo Scientific

OR480793-1G OR480793-5G
APOSOR480793-1GEA 180 EUR
APOSOR480793-1G APOSOR480793-5G
4-Bromo-3-methylthiophene-2-carboxaldehyde 97%
4-Bromo-3-methylthiophene-2-carboxaldehyde
Formel: C₆H₅BrOS
Lagertemperatur: Raumtemperatur
MDL: MFCD01859870
CAS-Nummer: 30153-47-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR