5-Chloro-1H-pyrrolo[3,2-b]pyridine 95%
Lieferant: Apollo Scientific
OR480796-5G
APOSOR480796-5GEA
178
EUR
APOSOR480796-5G
5-Chloro-1H-pyrrolo[3,2-b]pyridine 95%
5-Chloro-1H-pyrrolo[3,2-b]pyridine
Formel:
C₇H₅ClN₂ Molecular Weight: 152.5800 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08062553 CAS-Nummer: 65156-94-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...