(4S)-4-Methyloxazolidine-2,5-dione

Lieferant: Apollo Scientific

Synonyme: (S)-4-Methyloxazolidine-2,5-dione

OR480799-5G
APOSOR480799-5GEA 88.6 EUR
APOSOR480799-5G
(4S)-4-Methyloxazolidine-2,5-dione
(4S)-4-Methyloxazolidine-2,5-dione
Formel: C₄H₅NO₃
Molecular Weight: 115,087 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03411276
CAS-Nummer: 2224-52-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR