4-Isochinolinol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Isoquinolin-4-ol, 4-Hydroxyisoquinoline
OR48080-1G
OR48080-250MG
OR48080-5G
APOSOR48080-1GEA
88.6
EUR
APOSOR48080-1G
APOSOR48080-250MG
APOSOR48080-5G
4-Isochinolinol
4-Isochinolinol
Formel:
C₉H₇NO Molecular Weight: 145.1580 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00234514 CAS-Nummer: 3336-49-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...