1H-Pyrrolo[2,3-b]pyridin-4-ol hydrate 97%
Lieferant: Apollo Scientific
Synonyme:
1,7-dihydropyrrolo[2,3-b]pyridin-4-one, 4-Hydroxy-7-azaindole hydrate
OR480800-5G
APOSOR480800-5GEA
98.6
EUR
APOSOR480800-5G
1H-Pyrrolo[2,3-b]pyridin-4-ol hydrate 97%
1H-Pyrrolo[2,3-b]pyridin-4-ol hydrate
Formel:
C₇H₆N₂O·H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD08272229 CAS-Nummer: 74420-02-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...