2-Hydroxy-4-iodo-benzaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 2-Hydroxy-4-iodobenzaldehyde

OR480802-1G
APOSOR480802-1GEA 134 EUR
APOSOR480802-1G
2-Hydroxy-4-iodo-benzaldehyde 95%
2-Hydroxy-4-iodo-benzaldehyde
Formel: C₇H₅IO₂
Molecular Weight: 248.018 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08457196
CAS-Nummer: 38170-02-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR