4-Methoxy-1,5-dihydro-2H-pyrrol-2-one 98%

Lieferant: Apollo Scientific

Synonyme: 4-Methoxy-3-pyrrolin-2-one, 4-Methoxy-1H-pyrrol-2(5H)-one

OR480807-5G
APOSOR480807-5GEA 63.8 EUR
APOSOR480807-5G
4-Methoxy-1,5-dihydro-2H-pyrrol-2-one 98%
4-Methoxy-1,5-dihydro-2H-pyrrol-2-one
Formel: C₅H₇NO₂
Molecular Weight: 113,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00071564
CAS-Nummer: 69778-83-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR