Methylbenzyl isocyanide 95%

Lieferant: Apollo Scientific

OR480808-1G
APOSOR480808-1GEA 339 EUR
APOSOR480808-1G
Methylbenzyl isocyanide 95%
Methylbenzyl isocyanide
Formel: C₉H₉N
Molecular Weight: 131,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02664589
CAS-Nummer: 17329-20-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR