1H-Benzimidazole-2-carboxylic acid methyl ester 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 1H-benzo[d]imidazole-2-carboxylate

OR480814-5G
APOSOR480814-5GEA 140 EUR
APOSOR480814-5G
1H-Benzimidazole-2-carboxylic acid methyl ester 95%
1H-Benzimidazole-2-carboxylic acid methyl ester
Formel: C₉H₈N₂O₂
Molecular Weight: 176.172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03413579
CAS-Nummer: 5805-53-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR