(5S)-5-Aminopiperidin-2-one hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR480824-1G
APOSOR480824-1GEA 654 EUR
APOSOR480824-1G
(5S)-5-Aminopiperidin-2-one hydrochloride 97%
(5S)-5-Aminopiperidin-2-one hydrochloride
Formel: C₅H₁₀N₂O·ClH
Molecular Weight: 150,607 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03094719
CAS-Nummer: 672883-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR