Methyl 2-bromo-4-(bromomethyl)benzoate 97%

Lieferant: Apollo Scientific

OR480825-5G
APOSOR480825-5GEA 0 EUR
APOSOR480825-5G
Methyl 2-bromo-4-(bromomethyl)benzoate 97%
Methyl 2-bromo-4-(bromomethyl)benzoate
Formel: C₉H₈Br₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03840738
CAS-Nummer: 128577-48-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR