1-(2-Methoxyethyl)homopiperazine 95%

Lieferant: Apollo Scientific

Synonyme: 1-(2-Methoxyethyl)-1,4-diazepane

OR480833-1G
APOSOR480833-1GEA 150 EUR
APOSOR480833-1G
1-(2-Methoxyethyl)homopiperazine 95%
1-(2-Methoxyethyl)homopiperazine
Formel: C₈H₁₈N₂O
Molecular Weight: 158,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09054770
CAS-Nummer: 927802-38-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR