tert-Butyl (3R)-3-iodopyrrolidine-1-carboxylate 95%

Lieferant: Apollo Scientific

OR480835-1G
APOSOR480835-1GEA 294 EUR
APOSOR480835-1G
tert-Butyl (3R)-3-iodopyrrolidine-1-carboxylate 95%
tert-Butyl (3R)-3-iodopyrrolidine-1-carboxylate
Formel: C₉H₁₆INO₂
Molecular Weight: 297,133 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17015908
CAS-Nummer: 1234576-86-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR