4-Bromo-2-(methylthio)pyrimidine 97%

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-2-methylsulphanyl-pyrimidine

OR480843-5G
APOSOR480843-5GEA 77 EUR
APOSOR480843-5G
4-Bromo-2-(methylthio)pyrimidine 97%
4-Bromo-2-(methylthio)pyrimidine
Formel: C₅H₅BrN₂S
Molecular Weight: 205.07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878932
CAS-Nummer: 959236-97-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR