4-(1,3-Dioxo-1,3-dihydro-isoindol-2-yl)-2-oxo-butyric acid methyl ester 90%, tech

Lieferant: Apollo Scientific

OR480847-1G
APOSOR480847-1GEA 360 EUR
APOSOR480847-1G
4-(1,3-Dioxo-1,3-dihydro-isoindol-2-yl)-2-oxo-butyric acid methyl ester 90%, tech
4-(1,3-Dioxo-1,3-dihydro-isoindol-2-yl)-2-oxo-butyric acid methyl ester
Formel: C₁₃H₁₁NO₅
Molecular Weight: 261,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18434518
CAS-Nummer: 62987-16-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR