Ethyl 2-methyl-5-nitro-pyrazole-3-carboxylate 95%
Lieferant: Apollo Scientific
OR480849-1G
APOSOR480849-1GEA
440
EUR
APOSOR480849-1G
Ethyl 2-methyl-5-nitro-pyrazole-3-carboxylate 95%
Ethyl 2-methyl-5-nitro-pyrazole-3-carboxylate
Formel:
C₇H₉N₃O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09701484 CAS-Nummer: 1245807-07-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...