3-Amino-2-methoxypyridine-4-carboxylic acid 95%

Lieferant: Apollo Scientific

Synonyme: 3-Amino-2-methoxy-4-pyridinecarboxylic acid

OR480850-5G
APOSOR480850-5GEA 360 EUR
APOSOR480850-5G
3-Amino-2-methoxypyridine-4-carboxylic acid 95%
3-Amino-2-methoxypyridine-4-carboxylic acid
Formel: C₇H₈N₂O₃
Molecular Weight: 168,152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18262266
CAS-Nummer: 870997-81-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR