3-(Bromomethyl)-2-methylpyridine hydrobromide 95%

Lieferant: Apollo Scientific

OR480851-1G OR480851-500MG
APOSOR480851-1GEA 563 EUR
APOSOR480851-1G APOSOR480851-500MG
3-(Bromomethyl)-2-methylpyridine hydrobromide 95%
3-(Bromomethyl)-2-methylpyridine hydrobromide
Formel: C₇H₈BrN·BrH
Molecular Weight: 266.961 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22200150
CAS-Nummer: 76915-53-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR