3-(Bromomethyl)-2-methylpyridine hydrobromide 95%
Lieferant: Apollo Scientific
OR480851-1G
OR480851-500MG
APOSOR480851-1GEA
563
EUR
APOSOR480851-1G
APOSOR480851-500MG
3-(Bromomethyl)-2-methylpyridine hydrobromide 95%
3-(Bromomethyl)-2-methylpyridine hydrobromide
Formel:
C₇H₈BrN·BrH Molecular Weight: 266.961 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22200150 CAS-Nummer: 76915-53-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...