3-(3-Hydroxyphenyl)-piperazine-1-carboxylic acid tert-butyl ester 95%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 3-(3-hydroxyphenyl)piperazine-1-carboxylate

OR480858-1G OR480858-250MG
APOSOR480858-1GEA 1500 EUR
APOSOR480858-1G APOSOR480858-250MG
3-(3-Hydroxyphenyl)-piperazine-1-carboxylic acid tert-butyl ester 95%
3-(3-Hydroxyphenyl)-piperazine-1-carboxylic acid tert-butyl ester
Formel: C₁₅H₂₂N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD07021282
CAS-Nummer: 889956-76-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR