3-Chloro-7,8-dihydroquinolin-5(6H)-one 95+%

Lieferant: Apollo Scientific

OR480859-250MG
APOSOR480859-250MGEA 88.6 EUR
APOSOR480859-250MG
3-Chloro-7,8-dihydroquinolin-5(6H)-one 95+%
3-Chloro-7,8-dihydroquinolin-5(6H)-one
Formel: C₉H₈ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD10696330
CAS-Nummer: 127724-75-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR