8-Chloro-6-methoxyquinoline

Lieferant: Apollo Scientific

OR480860-1G
APOSOR480860-1GEA 0 EUR
APOSOR480860-1G
8-Chloro-6-methoxyquinoline
8-Chloro-6-methoxyquinoline
Formel: C₁₀H₈ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD09907835
CAS-Nummer: 796851-15-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR