4-Bromo-2,2'-bipyridyl 95%
Lieferant: Apollo Scientific
Synonyme:
4-Bromo-2,2'-bipyridine
OR480861-1G
APOSOR480861-1GEA
179
EUR
APOSOR480861-1G
4-Bromo-2,2'-bipyridyl 95%
4-Bromo-2,2'-bipyridyl
Formel:
C₁₀H₇BrN₂ Molecular Weight: 235,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09910216 CAS-Nummer: 14162-95-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...