2-(Chloromethyl)-3-methylpyridine hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR480862-1G
APOSOR480862-1GEA 127 EUR
APOSOR480862-1G
2-(Chloromethyl)-3-methylpyridine hydrochloride 95%
2-(Chloromethyl)-3-methylpyridine hydrochloride
Formel: C₇H₈ClN·ClH
Molecular Weight: 178,059 g/mol
Schmelzpunkt: 159 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08669778
CAS-Nummer: 4370-22-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR