3,4-Dibromofuran-2,5-dione 95%

Lieferant: Apollo Scientific

OR480863-1G
APOSOR480863-1GEA 119 EUR
APOSOR480863-1G
3,4-Dibromofuran-2,5-dione 95%
3,4-Dibromofuran-2,5-dione
Formel: C₄Br₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00030673
CAS-Nummer: 1122-12-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR