Methyl (2S)-2-amino-3-(4-iodophenyl)propanoate hydrochloride 95%

Lieferant: Apollo Scientific

OR480866-1G
APOSOR480866-1GEA 226 EUR
APOSOR480866-1G
Methyl (2S)-2-amino-3-(4-iodophenyl)propanoate hydrochloride 95%
Methyl (2S)-2-amino-3-(4-iodophenyl)propanoate hydrochloride
Formel: C₁₀H₁₂INO₂·ClH
Molecular Weight: 341,573 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08058398
CAS-Nummer: 113850-77-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR