(3R)-3-Methylpyrrolidine hydrochloride

Lieferant: Apollo Scientific

Synonyme: (R)-3-Methylpyrrolidine hydrochloride

OR480867-1G
APOSOR480867-1GEA 294 EUR
APOSOR480867-1G
(3R)-3-Methylpyrrolidine hydrochloride
(3R)-3-Methylpyrrolidine hydrochloride
Formel: C₅H₁₁N·ClH
Molecular Weight: 121,608 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06796631
CAS-Nummer: 235093-98-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR